D0252
4,6-Diamino-2-hydroxypyrimidine hemisulfate salt
Synonym(s):
6-Aminocytosine hemisulfate salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C4H6N4O · 1/2H2SO4
CAS Number:
Molecular Weight:
175.16
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
solubility
aqueous buffer: soluble
storage temp.
room temp
SMILES string
OS(O)(=O)=O.Nc1cc(N)nc(O)n1.Nc2cc(N)nc(O)n2
InChI
1S/2C4H6N4O.H2O4S/c2*5-2-1-3(6)8-4(9)7-2;1-5(2,3)4/h2*1H,(H5,5,6,7,8,9);(H2,1,2,3,4)
InChI key
ZLSLDSHQBJEJBG-UHFFFAOYSA-N
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service