Skip to Content
Merck
All Photos(1)

Key Documents

CPS350

Sigma-Aldrich

Chemiluminescent Peroxidase Substrate-3

sufficient for 50 mL substrate

Synonym(s):

CPS3

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
CHF 104.00

CHF 104.00


Check Cart for Availability


Select a Size

Change View
1 KIT
CHF 104.00

About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.83

CHF 104.00


Check Cart for Availability

form

solution

usage

sufficient for 50 mL substrate

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

CPS3 formulations are economically priced and outperform the most commonly used competitors′ products. This two component system provides low nanogram detection with superior signal-to-noise ratio. In addition, the components are stable after mixing, adding to the flexibility of usage.
Chemiluminescent Peroxidase Substrate-3 has been used for the quantification of aptamer binding.[1][2] It has also been used in endopeptidase assay to determine the enzymatic activity of botulinum neurotoxins (BoNTs).[3]

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High level expression and immunochemical characterization of botulinum neurotoxin type F light chain
Chauhan R, et al.
Protein Expression and Purification, 146, 51-60 (2018)
Selection of DNA aptamers for extra cellular domain of human epidermal growth factor receptor 2 to detect HER2 positive carcinomas
Sett A, et al.
Clinical And Translational Oncology, 19(8), 976-988 (2017)
Arghya Sett et al.
Translational research : the journal of laboratory and clinical medicine, 183, 104-120 (2017-01-15)
Estrogen receptor alpha (ERα) also known as NR3A1 (nuclear receptor subfamily 3, group A, member 1) is a ligand-activated transcription factor. It is an important biomarker for breast cancer metastasis. In the present study, we report a novel DNA aptamer
Yanpei Liu et al.
Plant physiology, 183(1), 194-205 (2020-03-12)
Photorespiration is an essential process in oxygenic photosynthetic organisms triggered by the oxygenase activity of Rubisco. In peroxisomes, photorespiratory HYDROXYPYRUVATE REDUCTASE1 (HPR1) catalyzes the conversion of hydroxypyruvate to glycerate together with the oxidation of a pyridine nucleotide cofactor. HPR1 regulation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service