Documents
CLS9186
Corning® aspiration pipettes
volume 2 mL
Synonym(s):
Aspiration pipette, Corning pipette
About This Item
UNSPSC Code:
41121500
eCl@ss:
32070201
NACRES:
NB.01
material
polystyrene
sterility
sterile
packaging
pack of 100 ea
case of 1000 ea
manufacturer/tradename
Corning 9186
capacity
2 mL
volume
2 mL
General description
  • Corning® aspirating pipettes are made of optically clear polystyrene for durability and good mechanical strength.
  • The sterile, ungraduated, and unplugged pipettes are best suited for aspirating liquid aseptically using vacuum suction.
  • Individually wrapped and bulk packed.
  • Disposable and certified non-pyrogenic.
Aspirating pipettes are sterile, ungraduated and unplugged polystyrene pipettes for aspirating liquid using vacuum suction.
Legal Information
Corning is a registered trademark of Corning, Inc.