CLS4801
Corning® Isotip® filtered pipet tips
0.1-2.0 μL, universal fit racked pipet tips, natural, sterile, 96 tips/rack, 960 tips/case
Synonym(s):
pipet tips, pipette tips
About This Item
Recommended Products
material
natural tip
polypropylene
sterility
sterile
feature
DNase free
RNase free
barrier: no
disposable
endotoxin free
filter
graduation
packaging: rack
packaging
rack of 96
manufacturer/tradename
Corning 4801
volume range
0.1-2 μL
suitability
suitable for (applications where avoidance of cross contamination is critical, such as DNA amplification and radioisotope handling)
compatibility
all popular research-grade pipettors
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Corning Isotip filtered pipet tips feature an inert, hydrophobic barrier that prevents aerosolised contaminants from contacting pipettor shafts. All filtered pipet tips are packaged sterile and are certified RNase/DNase-free and nonpyrogenic.
Application
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service