CLS4487LE
Corning® Costar® Stripette® serological pipettes, individually paper/plastic wrapped
capacity 5 mL, non-color coded, pkg of 200 ea
Synonym(s):
pipet
About This Item
material
polystyrene
description
available in Asia only.
sterility
sterile
feature
disposable
packaging
pkg of 200 ea
manufacturer/tradename
Corning 4487LE
measuring range
accuracy: 0.1 mL
capacity
5 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Color-coding on paper allows easy size identification.
- Paper/plastic wrap allows full view
- Unique color-coded magnifying stripe for easier meniscus viewing.
- Sterile, individually wrapped and certified nonpyrogenic and DNase-/RNase-free
- Accuracy within ± 2% at full volume
- Lot numbers printed on each pipet wrapper
- Calibrated to deliver (TD-EX 20ºC) their contents with blowout
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service