CLS432108
Corning® Ice Pan, rectangular
midi, capacity 4 L, pink
Synonym(s):
Corning storage container
Select a Size
CHF 131.00
Select a Size
About This Item
CHF 131.00
Recommended Products
feature
midi
packaging
pkg of 1
manufacturer/tradename
Corning 432108
L × W × H
31.1 cm × 22.2 cm × 11.4 cm
color
pink
Looking for similar products? Visit Product Comparison Guide
General description
- Non-toxic, recyclable ethyl-vinyl acetate (EVA) foam containers for use with ice, dry ice, liquid nitrogen, alcohol slurries.
- Will not sweat, leak or skid on bench.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service