Skip to Content
Merck
All Photos(1)

Key Documents

CLS3826BC

Corning® 384 well plates, low volume

size 384 well, polystyrene, flat bottom white polystyrene wells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

material

flat bottom white polystyrene wells
polystyrene

sterility

sterile

feature

lid

packaging

case of 50 ea

manufacturer/tradename

Corning 3826BC

maximum volume

50 μL

size

384 well
384 wells

working volume

40 μL

color

white

suitability

suitable for (immunoassay; liquid-handling procedures)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Corning® 384 Well Plates, Low Volume

The 384 multiwell low volume plate dimensions conform to proposed SBS standards. The wells for each microplate have either a square to round design accompanied by a flat bottom. In addition, each well has a raised rim to reduce cross contamination during assay and liquid-handling process.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giacomo Pirovano et al.
European journal of nuclear medicine and molecular imaging, 47(4), 1003-1010 (2019-11-18)
OTS514 is a highly specific inhibitor targeting lymphokine-activated killer T cell-originated protein kinase (TOPK). A fluorescently labeled TOPK inhibitor could be used for tumor delineation or intraoperative imaging, potentially improving patient care. Fluorescently labeled OTS514 was obtained by conjugating the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service