Key Documents
CLS3809
Corning® 96 Well CellBIND® Microplates
white polystyrene plate, sterile, flat bottom clear wells, lid, case of 100 plates
Synonym(s):
cell culture plate, multi well plates, multiple well plates
Select a Size
100 EA
CHF 1’580.00
CHF 1’580.00
Check Cart for Availability
Select a Size
Change View
100 EA
CHF 1’580.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
material:
flat bottom clear wells
flat bottom wells
polystyrene plate
white polystyrene plate
size:
96 wells
CHF 1’580.00
Check Cart for Availability
material
flat bottom clear wells
flat bottom wells
polystyrene plate
white polystyrene plate
sterility
sterile
sterile
feature
lid
lid
plate format: 96 well standard
packaging
case of 100 plates
pack of 5
case of 50
manufacturer/tradename
Corning 3809
size
96 wells
surface area
0.32 cm2 , cell growth area
cell growth area
cell growth area
well volume
360 μL
well working volume
75-200 μL
suitability
suitable for (cell cuture)
General description
  • Corning® 96 Well CellBIND® Microplates have an optically clear flat bottom for direct microscopic viewing.
  • Thinner bottom enables lower background fluorescence and allows readings down to 340 nm.
  • Opaque walls aid in minimal well-to-well cross-talk.
  • Flat bottoms with 360 μL total volume
  • Recommended working volumes of 75 to 200 μL
  • CellBIND® surface treated for optimal cell attachment
  • Nonreversable lids with condensation rings to reduce contamination
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Individual alphanumeric codes for well identification
Legal Information
CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)