CLS353104
Corning® Falcon® Permeable Support
for use with 24 Well Plate, with 1.0 µm Transparent PET Membrane, sterile
Synonym(s):
Permeable Support
About This Item
sterility
sterile
feature
with 1.0 µm Transparent PET Membrane
packaging
case of 48 ea (Individually wrapped)
manufacturer/tradename
Corning 353104
compatibility
for use with 24 Well Plate
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service