Key Documents
CLS3368
Corning® 96 Well EIA/RIA Assay Microplate
flat bottom clear, polystyrene, bag of 25, sterile, lid: no
Synonym(s):
96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 welll plate, EIA/RIA plate, immunoassay plate
Select a Size
100 EA
CHF 408.00
CHF 408.00
Check Cart for Availability
Select a Size
Change View
100 EA
CHF 408.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
material:
flat bottom clear
polystyrene
polystyrene
round wells
size:
96 wells , 96 well plate
CHF 408.00
Check Cart for Availability
material
flat bottom clear
polystyrene
polystyrene
round wells
sterility
sterile
feature
lid: no
skirt
plate format: 96 well Easy-Wash
well: flat bottom, clear
packaging
case of 100
bag of 25
manufacturer/tradename
Corning 3368
size
96 wells , 96 well plate
well volume
330-360 μL , Note: CLS3366 and CLS3797 have well volume capacities of 330 uL
well working volume
75-200 μL
suitability
suitable for (EIA/RIA; immunoassays)
binding type
non-treated surface
General description
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
  • Is suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
  • Untreated microplates have a binding capacity of approximately 100 to 200ng IgG/cm2
  • Easy Wash design (round to narrowed flat well bottom) for improved washing in immunoassays
  • EIA/RIA plates are tested on lot-to-lot basis and the certification results for each lot are available upon request
  • Recommended working volume of up to 75 to 200 μL
  • Conforms to standard microplate footprint and dimensions
These 96 well EIA/RIA plates are designed for in vitro diagnostic immunoassays. All EIA/RIA plates are manufactured from special medical-grade polystyrene for uniform binding, high optical clarity and low background absorption. In addition, the Easy-Wash products are part of the Certified Surface Chemistry Programme, which guarantees protein binding performance to be within QA release specifications.
Legal Information
Corning is a registered trademark of Corning, Inc.
Easy-Wash is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)