Skip to Content
Merck
All Photos(1)

Key Documents

CLS2592

Corning® 1 x 8 Stripwell 96 well plates

high binding surface, clear

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
CHF 245.00
100 EA
CHF 701.00

CHF 245.00


Estimated to ship on03 April 2025



Select a Size

Change View
25 EA
CHF 245.00
100 EA
CHF 701.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
flat bottom clear polystyrene (96-well)
round wells
size:
96 wells
well volume:
360 μL
well working volume:
75-200 μL , for low volume plates volume is 50-125 μL

CHF 245.00


Estimated to ship on03 April 2025


material

flat bottom clear polystyrene (96-well)
round wells

sterility

non-sterile

feature

lid: no
skirt

packaging

plate of 12 strips

manufacturer/tradename

Corning 2592

size

96 wells

well volume

360 μL

well working volume

75-200 μL , for low volume plates volume is 50-125 μL

color

clear

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

The Costar 1 × 8 strips are pre-assembled in an "egg crate" style 96 well strip holder. Strips fit in holder only one way, eliminating mis-orientation. If covers are needed, the notched cover fits the fully assembled strip holder.

Features and Benefits

  • Corning® Stripwell 96 well plates are designed for a variety of laboratory assays like immunoassays.
  • High binding hydrophobic surface binds medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic region.

Legal Information

Corning is a registered trademark of Corning, Inc.
Stripwell is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pradeep Ramalingam et al.
Nature communications, 11(1), 666-666 (2020-02-06)
Inflammatory signals arising from the microenvironment have emerged as critical regulators of hematopoietic stem cell (HSC) function during diverse processes including embryonic development, infectious diseases, and myelosuppressive injuries caused by irradiation and chemotherapy. However, the contributions of cellular subsets within

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service