Skip to Content
Merck
All Photos(1)

Key Documents

fR2

98031102, human breast, Epithelial, cuboidal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

fR2, 98031102

biological source

human breast

growth mode

Adherent

karyotype

Aneuploid

morphology

Epithelial, cuboidal

shipped in

dry ice

storage temp.

−196°C

General description

For complete product information, please see fR2

Cell Line Origin

Human breast epithelial, SV40 transformed

Cell Line Description

The SV40 transformed breast epithelial cell line fR2 was obtained in 1982 by infecting suspensions of primary milk cultures with wild-type SV40. The cells have been shown to express Muc-1, also known as HMFG-1 antigen, which is a mucin-like component of human milk fat globule membranes. They are positive for keratin 8 and 18, but negative for keratins 4, 6, 7, 10, 13, 14, 16, 17 and 19. Although fR2 is morphologically different from fR5 (Sigma Catalogue no. 98031103), they have probably been derived from a common precursor cell since the karyotypes are similar. Karyotype analysis revealed hypotetraploidy and several rearrangements involving chromosome 1 and 11 which are frequently found in breast carcinomas and lines derived from metastatic pleural effusions. fR2 cells display anchorage independent growth in soft agar and are positive for SV40 T-antigen. Early passages were non-tumorigenic in nude mice.

Application

Study of human mammary epithelial cells

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 12,13
D13S317: 12
D16S539: 11,13
D5S818: 12,13
D7S820: 8,12
THO1: 9.3
TPOX: 8
vWA: 20

Culture Medium

RPMI 1640 + 2mM Glutamine + 10μg/ml insulin + 5μg/ml hydrocortisone + 10% FBS

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 i.e. seeding 1 x 10,000 cells / cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service