Skip to Content
Merck
All Photos(1)

Documents

PMK Cell Line macaque

98020308, rhesus macaque kidney

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

PMK Cell Line macaque, 98020308

biological source

rhesus macaque kidney

description

Primary rhesus macaque kidney cells, blood, serum

shipped in

dry ice

storage temp.

−196°C

General description

Tested for Herpes B, Hepatitis A & B, SIV, STLV, Type D retrovirus, Salmonella, Shigellla, Helminths and Entomoeba. This cell line can be supplied on a standing order basis, please contact ECACC for further details.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service