AFRC MAC 160
Reactivity: Anti-Cytokinin, plant, 93091001
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
Mouse x Rat Hybridoma
ImmunoDon
F344 rat spleen
Myeloma
NS0
Reactivity
Anti-Cytokinin, plant
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse NS0 x Rat F344 spleen
Cell Line Description
The antibody is directed against a range of plant cytokinins including isopentenyladenosine, isopentenyladenine and benzyladenine riboside.
Immunogen
Isopentenyladenosine
Application
Immunoaffinity chromatography
The isotype of the monoclonal antibody produced by this cell line is rat IgG2a.
Culture Medium
DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service