Skip to Content
Merck
All Photos(1)

Documents

U-2 OS Cell Line human

92022711, human bone, Not specified

Synonym(s):

U-2OS Cells, U2-OS Cells, U20S Cells, U2OS Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

U-2 OS Cell Line human, osteosarcoma, 92022711

biological source

human bone

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human Osteosarcoma

Cell Line Description

Cell line derived in 1964 from a moderately differentiated sarcoma of the tibia of a 15 year old girl.

Application

U-2 OS has been used to study:
  • the importance of cyclin D1 for the activity of lithocholic acid hydroxyamide (LCAHA)
  • the interaction of human single-stranded DNA binding protein 1 (hSSB1) with bloom syndrome protein helicase (BLM helicase)
  • calcium-mediated actin reset (CaAR) in response to physiological changes

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 13
D13S317: 13
D16S539: 11,12
D5S818: 11
D7S820: 11,12
THO1: 6,9.3
TPOX: 11,12
vWA: 14,18

Culture Medium

McCoy′s 5a medium with 1.5 mM Glutamine; 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split subconfluent 0.25% trypsin, 0.03% EDTA solution. Remove the solution and add an additional 1 to 2 ml of trypsin-EDTA solution. Allow the flask to sit at room temperature (or at 37°C) until the cells detach. Add fresh medium, aspirate and dispense into new culture flasks.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cell cycle regulates vital processes like DNA repair, cancer prevention. Four stages: G1, S, G2, M. NTPs don't permeate membranes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service