Skip to Content
Merck
All Photos(1)

Documents

H139-52

Reactivity: Anti-κ-Chain, mouse, 85103111

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

Mouse x Rat Heterohybridoma

ImmunoDon

LOU/Ws1 rat spleen

Myeloma

P3X63Ag8.653

Reactivity

Anti-κ-Chain, mouse

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse P3X63Ag8.653 x Rat LOU/Ws1 spleen

Cell Line Description

The antibody is directed against murine kappa chain.

Immunogen

Purified murine immunoglobulin

Application

The isotype of the monoclonal antibody produced by this cell line is rat IgG1, kappa.

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. These cells require mouse macrophage feeder layers upon resuscitation

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service