Skip to Content
Merck
All Photos(1)

Key Documents

348C6

Reactivity: Anti-F5 (K99) pilus antigen of E.coli, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 85062511

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Bovine Heterohybridoma

packaging

tube of 5 μg 85062511-DNA-5UG
pkg of vial of cells 85062511-1VL

ImmunoDon

Jersey calf lymph node

Myeloma

94A1 (mouse x calf x calf 2° hybridoma)

Reactivity

Anti-F5 (K99) pilus antigen of E.coli

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse x Bovine 94A1 x Jersey calf lymph node

Cell Line Description

The antibody is directed against the F5 (K99) pilus antigen of E. coli.

Immunogen

Purified pilus antigen of F5 (K99+) enteroxigenic E.coli

Application

The isotype of the monoclonal antibody produced by this cell line is bovine IgG1.

Culture Medium

RPMI 1640 + 2mM glutamine + 1mM Sodium Pyruvate (NaP) + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service