Skip to Content
Merck
All Photos(1)

Documents

htr-20

Mouse monoclonal against human CD-120a, 10080903

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Human x Mouse Hybridoma

ImmunoDon

Mouse (Mus musculus) spleen

Myeloma

PAI

Reactivity

Human CD120a (TNF receptor p55)

technique(s)

cell culture | mammalian: suitable

isotype

IgG2b

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Not specified

Immunogen

Human CD120a (TNF receptor p55)

Application

Sandwich ELISA for soluble p55 TNF-R.

Culture Medium

Iscoves Modified Dulbeccos Medium (IMDM) + 2 mM glutamine + 25 U/ml Interleukin-6 (IL-6) + 5-10% Fetal Bovine Serum (FBS). Where the cells are grown under static conditions, 0.02% EDTA maybe required to dislodge them from the plastic.

Subculture Routine

Seed cultures between 1-3 x 105 cells/ml; Saturation density 8-10 x 105 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service