Skip to Content
Merck
All Photos(1)

Key Documents

293-Hektor

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

human kidney

packaging

tube of 5 μg 05030204-DNA-5UG
pkg of vial of cells 05030204-1VL

growth mode

Aggregates in suspension

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Embryo Kidney, serum-free

Cell Line Description

The human embryo kidney cell line 293 (Sigma Catalogue number. 85120602) adapted to grow in the chemically defined protein- and peptide-free Hektor G medium. Cells grow in suspension and tend to form aggregates in static culture. The cells grow as finely dispersed single cell suspension under optimal agitated culture conditions.

Culture Medium

Hektor G medium + 4mM Glutamine + 5 ug/ml Insulin. The exogeneous insulin supplementation can be reduced or totally eliminated under optimal culture conditions. When freezing cells down use culture medium (50:50 conditioned medium:fresh medium) + 10% DMSO.

Subculture Routine

Viability may be poor on resuscitation and may initially decrease further. Full recovery may take up to 2 weeks. A centrifugation step to remove the cryoprotectant is essential. Rapidly thaw the frozen ampoule in a water bath at 37°C for 1-2 minutes. Transfer the contents to a centrifuge tube and slowly add 5-10ml of pre-warmed growth media. Remove a sample for counting. Centrifuge at 100g for 2-3 minutes to pellet cells and seed at a relatively high density of 5-7 x 105 cells/ml. Leave culture flask upright and observe regularly until viable proliferating cell clumps form. Once established use a split ratio of 1:2 approximately every 4 to 5 days; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Legal Information

Hektor is a trademark of Cell Culture Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service