Skip to Content
Merck
All Photos(1)

Key Documents

C8977

Sigma-Aldrich

Monoclonal Anti-CREB antibody produced in rabbit

clone 48H2, purified immunoglobulin, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 ML
CHF 591.00

CHF 591.00


Check Cart for Availability


Select a Size

Change View
0.1 ML
CHF 591.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

CHF 591.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

48H2, monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen 43 kDa

species reactivity

rat, human, monkey, mouse

technique(s)

ChIP: 1:50
flow cytometry: 1:400
immunocytochemistry: 1:800
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:6,400
immunohistochemistry (frozen sections): 1:6,400
immunoprecipitation (IP): 1:250
indirect immunofluorescence: 1:800
western blot: 1:1,000 using extracts from SK-N-MC cells treated with forskolin or FGF

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CREB1(1385)
mouse ... Creb1(12912)
rat ... Creb1(81646)

Related Categories

Specificity

Detects endogenous levels of total CREB protein.

Immunogen

Recombinant protein specific to amino terminus of human CREB.

Physical form

Solution in 10 mM sodium HEPES, pH 7.5, containing 150 mM sodium chloride, 100 μg/mL bovine serum albumin, and < 0.02% sodium azide with 50% glycerol.

Legal Information

Rabbit monoclonals produced using Epitomics® Technology, U.S.Patent No. 5,675,063.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service