Skip to Content
Merck
All Photos(1)

Key Documents

C2490

Sigma-Aldrich

CYN 154806 trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

Cyanamid 154806 trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
CHF 199.00

CHF 199.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
500 μG
CHF 199.00

About This Item

Empirical Formula (Hill Notation):
C56H68N12O14S2 · xC2HF3O2
CAS Number:
Molecular Weight:
1197.34 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

CHF 199.00


Estimated to ship on02 June 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

lyophilized powder (solid)

color

white

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.C[C@@H](O)C1NC(=O)[C@H](CCCCN)NC(=O)[C@@H](Cc2c[nH]c3ccccc23)NC(=O)[C@H](Cc4ccc(O)cc4)NC(=O)[C@@H](CSSC[C@H](NC1=O)C(=O)N[C@H](Cc5ccc(O)cc5)C(N)=O)NC(=O)[C@H](Cc6ccc(cc6)[N+]([O-])=O)NC(C)=O

InChI

1S/C56H68N12O14S2.C2HF3O2/c1-30(69)48-56(80)66-47(54(78)62-42(49(58)73)23-33-12-18-37(71)19-13-33)29-84-83-28-46(65-51(75)43(60-31(2)70)24-32-10-16-36(17-11-32)68(81)82)55(79)63-44(25-34-14-20-38(72)21-15-34)52(76)64-45(26-35-27-59-40-8-4-3-7-39(35)40)53(77)61-41(50(74)67-48)9-5-6-22-57;3-2(4,5)1(6)7/h3-4,7-8,10-21,27,30,41-48,59,69,71-72H,5-6,9,22-26,28-29,57H2,1-2H3,(H2,58,73)(H,60,70)(H,61,77)(H,62,78)(H,63,79)(H,64,76)(H,65,75)(H,66,80)(H,67,74);(H,6,7)/t30-,41+,42-,43+,44+,45-,46-,47+,48+;/m1./s1

InChI key

FYVCFCQFOBEWIO-GVGJFGMUSA-N

Amino Acid Sequence

Ac-NO2-Phe-Cys-Tyr-Trp-Lys-Thr-Cys-Tyr-NH2

Biochem/physiol Actions

Potent and selective sst2 somatostatin receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caroline Nunn et al.
Naunyn-Schmiedeberg's archives of pharmacology, 367(1), 1-9 (2003-03-05)
The two forms (DTyr8 and LTyr8) of the putative somatostatin sst2 receptor antagonist CYN 154806 (Ac-4NO2-Phe-c(DCys-Tyr-DTrp-Lys-Thr-Cys)-D/LTyr-NH2) were investigated on recombinant human somatostatin receptors and endogenous guinea-pig ileum receptors. In radioligand binding studies using the agonist radioligands [125I]LTT-SRIF-28, [125I][Tyr10]cortistatin-14, [125I]CGP 23996
Jason P Hannon et al.
Journal of molecular neuroscience : MN, 18(1-2), 15-27 (2002-04-05)
Somatostatin (SRIF, somatotropin release inhibiting factor), discovered for its inhibitory action on growth hormone (GH) secretion from pituitary, is an abundant neuropeptide. Two forms, SRIF14 and SRIF28 exist. Recently, a second family of peptides with very similar sequences and features
Massimo Dal Monte et al.
Naunyn-Schmiedeberg's archives of pharmacology, 367(2), 188-192 (2003-02-22)
In the mammalian retina, somatostatin (SRIF-14) acts through distinct receptor subtypes (sst(1-5)). Among them, sst(2) has been localized to numerous retinal cells, including photoreceptors and rod bipolar cells (RBCs). The specific role of sst(2) in the retina is largely undetermined.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service