Skip to Content
Merck
All Photos(1)

Documents

C1187

Sigma-Aldrich

Color Coders for Nalgene® cryogenic vials

white

Synonym(s):

vial cap inserts, vial identifier

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103032
NACRES:
NB.22

material

polystyrene

manufacturer/tradename

Nalgene®

color

white

Looking for similar products? Visit Product Comparison Guide

General description

Colored disks fit into the recessed tops of Nalgene cryogenic vials. The system allows color-coding of vials for quick visual identification. Cut out top facilitates handling of vials with forceps. Flat surface permits writing.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service