Skip to Content
Merck
All Photos(1)

Documents

C0921

Sigma-Aldrich

Silicone sponge closures

size 28 mm

Synonym(s):

Closures for Vials, Silicone Closures, Sponge Closures

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

size

28 mm

Looking for similar products? Visit Product Comparison Guide

General description

Silicone rubber rings with sealed-in silicone sponge filters.

These silicone sponge closures are ideal when working with aerobic cultures; they permit an efficient exchange of gases. They also cut losses due to evaporation in half in comparison to cotton plugs, thus minimizing changes in pH values. They can be dry heat sterilized repeatedly at 180°C and are easy to clean and maintain (reusable).

Compatibility

For use with 125-300 mL flasks.
Does not fit Nalgene polycarbonate shake flasks.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service