Skip to Content
Merck
All Photos(1)

Key Documents

C0358

Sigma-Aldrich

Z-Leu-Leu-Phe-CHO

≥95% (HPLC), solid

Synonym(s):

Cbz-Leu-Leu-Phe-CHO, Z-LLF-CHO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H39N3O5
CAS Number:
Molecular Weight:
509.64
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (HPLC)

form

solid

solubility

DMSO: soluble

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)OCc1ccccc1)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](Cc2ccccc2)C=O

InChI

1S/C29H39N3O5/c1-20(2)15-25(27(34)30-24(18-33)17-22-11-7-5-8-12-22)31-28(35)26(16-21(3)4)32-29(36)37-19-23-13-9-6-10-14-23/h5-14,18,20-21,24-26H,15-17,19H2,1-4H3,(H,30,34)(H,31,35)(H,32,36)/t24-,25-,26-/m0/s1

InChI key

LIOIGKLWXFXXEG-GSDHBNRESA-N

Amino Acid Sequence

Z-Leu-Leu-Phe-al

General description

Manufactured for Sigma by Boston Biochem., Inc.

Biochem/physiol Actions

Cell permeable proteasome inhibitor acting on the chymotrypsin-like activity of the multicatalytic proteinase complex.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S L Schauer et al.
Journal of immunology (Baltimore, Md. : 1950), 160(9), 4398-4405 (1998-05-09)
Recently, we demonstrated maintenance of nuclear factor (NF)-kappaB/Rel factors plays a major role in B cell survival. Treatment of WEHI 231 immature B cells with an Ab against the surface IgM protein (anti-IgM) induces apoptosis that can be rescued by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service