Skip to Content
Merck
All Photos(2)

Key Documents

BM991.1536.01

B Medical MT25

Transport Device (Box)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103025
Pricing and availability is not currently available.

material

polyethylene device

feature

CE compliant: no
CSA approved

packaging

pack of 1 ea

manufacturer/tradename

B Medical Systems

capacity

1.55 ft3

shipped in

ambient

storage temp.

room temp

General description

1.55 cu.ft., Passive, Transport Box, Capacity: (26) 450ml or (40) 270ml bag , Top Loading, Ext/Int: Molded Polyethylene, Cold Life: up to 109 hrs. At +32°C /up to 74 hrs. at +43°C

Application

Cold Chain

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service