Key Documents
BAF188510017
Bel-Art® Freezer Storage Box
no. of holes, 100, black polypropylene, pack of 5
Synonym(s):
cryopreservation storage rack, cryovial storage, indexed tube rack, microtube storage
About This Item
UNSPSC Code:
41123304
NACRES:
NB.22
material
black polypropylene
feature
for tubes 0.5, 1.5, and 2.0 mL
no. of holes 100
packaging
pack of 5
manufacturer/tradename
Bel-Art F18851-0017
General description
Sturdy freezer boxes provide compact storage and keep microcentrifuge tubes organized on the bench, in the freezer, or during transport.
Features and Benefits
  • Alphanumeric Indexing
  • Translucent lid for easy access and viewing; matte panels can be used for labeling (100-place boxes have hinged lid, 81-place boxes have lift off lid)
  • Withstand -80°C to +121°C (-112° to +249°F)
  • Stackable with molded-in ′no slide′ features
  • Assorted Colors with translucent lids (natural, blue, green, orange and purple)
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)