Skip to Content
Merck
All Photos(1)

Key Documents

B8685

Sigma-Aldrich

BM 15766 sulfate

≥98% (HPLC), powder

Synonym(s):

4-[2-[4-[3-(4-Chlorophenyl)-2-propenyl]-1-piperazinyl]ethyl]benzoic acid sulfate

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CHF 209.00
5 MG
CHF 792.00

CHF 209.00


Estimated to ship on09 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
CHF 209.00
5 MG
CHF 792.00

About This Item

Empirical Formula (Hill Notation):
C22H25ClN2O2 · H2SO4
CAS Number:
Molecular Weight:
482.98
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

CHF 209.00


Estimated to ship on09 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

mp

>300 °C

solubility

DMSO: soluble 4 mg/mL
water: insoluble

originator

Roche

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.OC(=O)c1ccc(CCN2CCN(CC2)CC=Cc3ccc(Cl)cc3)cc1

InChI

1S/C22H25ClN2O2.H2O4S/c23-21-9-5-18(6-10-21)2-1-12-24-14-16-25(17-15-24)13-11-19-3-7-20(8-4-19)22(26)27;1-5(2,3)4/h1-10H,11-17H2,(H,26,27);(H2,1,2,3,4)

InChI key

OVMQIVIYKXXEIT-UHFFFAOYSA-N

Biochem/physiol Actions

Dehydrocholesterol reductase inhibitor. Inhibits 7-dehydrocholesterol reductase, which catalyzes the last step of cholesterol synthesis.

Features and Benefits

This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cholesterol synthesis regulation by dietary levels, LDL receptors control lipid-rich LDL particle transport in cells.

Discover Bioactive Small Molecules for Lipid Signaling Research

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service