Skip to Content
Merck
All Photos(1)

Key Documents

B7753

Sigma-Aldrich

Butyl 4-aminobenzoate

Synonym(s):

Butamben

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CO2(CH2)3CH3
CAS Number:
Molecular Weight:
193.24
Beilstein:
1211465
EC Number:
MDL number:
UNSPSC Code:
12352100

bp

174 °C/8 mmHg (lit.)

mp

57-58 °C (lit.)

SMILES string

CCCCOC(=O)c1ccc(N)cc1

InChI

1S/C11H15NO2/c1-2-3-8-14-11(13)9-4-6-10(12)7-5-9/h4-7H,2-3,8,12H2,1H3

InChI key

IUWVALYLNVXWKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye Joo Kim et al.
Bioorganic & medicinal chemistry letters, 21(24), 7363-7366 (2011-11-02)
As the primary step for 'drug repositioning', we evaluated the effect of 2000 drugs and drug candidates on the commitment of bi-potential mesenchymal precursor C2C12 cells into osteoblasts in the presence of bone morphogenetic protein (BMP)-2 and found that butamben
Alfonso Pérez-Garrido et al.
Bioorganic & medicinal chemistry, 17(2), 896-904 (2008-12-06)
This paper reports a QSAR study for predicting the complexation of a large and heterogeneous variety of substances (233 organic compounds) with beta-cyclodextrins (beta-CDs). Several different theoretical molecular descriptors, calculated solely from the molecular structure of the compounds under investigation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service