Skip to Content
Merck
All Photos(1)

Documents

B7446

Sigma-Aldrich

Bovine Calf Serum

Australia origin, sterile-filtered, suitable for cell culture, suitable for hybridoma

Synonym(s):

Calf Serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207

biological source

bovine fetus

sterility

sterile-filtered

form

liquid

origin

Australia origin

technique(s)

cell culture | hybridoma: suitable
cell culture | mammalian: suitable

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Animal serum is commonly used to supplement basal media formulations for the optimal growth of many cell types in vitro.
Bovine Calf Serum (BCS) is collected from calves that are less than 12 months old. The serum contains approximately double the protein of Fetal Bovine Serum (FBS). BCS will promote growth in a variety of mammalian cell lines and is often used as a cost-effective substitute for FBS.

Storage and Stability

To effectively preserve the integrity of animal serum, it should be stored frozen and protected from light. Multiple thaw/freeze cycles should be avoided as they will hasten the degradation of serum nutrients and can result in the formation of insoluble precipitates.
For stability and optimal performance, serum should be stored at −20 °C and used prior to the labeled expiration date.

Analysis Note

Endotoxin and hemoglobin tested

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service