B7430
Nalgene® dropper bottle with screw closure
capacity 15 mL, white bottle, assorted colors cap
Synonym(s):
bottle dropper, dropper bottles
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
low-density polyethylene
round bottle
white bottle
Joint
Joints threaded neck (15-415)
packaging
pack of 25 ea
manufacturer/tradename
Nalgene® 2753-9050
capacity
15 mL
color
assorted colors cap
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
- Nalgene® dropper bottles with screw closure is an alternative to various pipetting devices.
- The tip ensures accurate dispensing of 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time.
- Chemically resistant to most reagents.
- LDPE clear transparent bottles for easy sample visualization.
- Leak-proof closure - prevents sample loss.
- Control dispensing tip ensures a secure fit.
- Suitable for use in biotech, diagnostic and pharmaceutical applications.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service