B5309
Lys-Lys-(Hyp3,β-(2-thienyl)-Ala5,8,D-Phe7)-Bradykinin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C62H95N19O14S2
Molecular Weight:
1394.67
MDL number:
UNSPSC Code:
12352200
composition
Peptide content, ≥70%
storage temp.
−20°C
SMILES string
NCCCC[C@H](N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N1CCC[C@H]1C(=O)N2C[C@H](O)C[C@H]2C(=O)NCC(=O)N[C@@H](Cc3cccs3)C(=O)N[C@@H](CO)C(=O)N[C@H](Cc4ccccc4)C(=O)N[C@@H](Cc5cccs5)C(=O)N[C@@H](CCCNC(N)=N)C(O)=O
Amino Acid Sequence
Lys-Lys-Arg-Pro-Hyp-Gly-β-(2-thienyl)-Ala-Ser-D-Phe-β-(2-thienyl)-Ala-Arg-OH
Biochem/physiol Actions
An analog of bradykinin which is a specific inhibitor of five different actions of bradykinin.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
T Griesbacher et al.
British journal of pharmacology, 92(2), 333-340 (1987-10-01)
1 The inhibition of the bradykinin-induced plasma extravasation by six bradykinin (Bk) antagonists was tested on rabbit skin. All of them showed inhibitory effects without an agonistic action in the does used. B4310 (Lys-Lys-3-Hyp-5,8-Thi-7-DPhe-Bk) was the most active antagonist and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service