Skip to Content
Merck
All Photos(1)

Key Documents

B5005

Sigma-Aldrich

Behenoyl chloride

≥99%

Synonym(s):

Docosanoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)20COCl
CAS Number:
Molecular Weight:
359.03
Beilstein:
1792888
MDL number:
UNSPSC Code:
12352211

Assay

≥99%

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCC(Cl)=O

InChI

1S/C22H43ClO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h2-21H2,1H3

InChI key

QTHQYNCAWSGBCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service