Skip to Content
Merck
All Photos(1)

Key Documents

B4438

Sigma-Aldrich

BVT.13 hydrate

≥98% (HPLC)

Synonym(s):

2-[(2,4-Dichlorobenzoyl)amino]-5-(2-pyrimidinyloxy)-benzoic acid hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H11Cl2N3O4 · xH2O
CAS Number:
Molecular Weight:
404.20 (anhydrous basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

off-white

solubility

DMSO: >5 mg/mL

storage temp.

2-8°C

SMILES string

O.OC(=O)c1cc(Oc2ncccn2)ccc1NC(=O)c3ccc(Cl)cc3Cl

InChI

1S/C18H11Cl2N3O4.H2O/c19-10-2-4-12(14(20)8-10)16(24)23-15-5-3-11(9-13(15)17(25)26)27-18-21-6-1-7-22-18;/h1-9H,(H,23,24)(H,25,26);1H2

InChI key

FMWLWXPXWXDYMJ-UHFFFAOYSA-N

Biochem/physiol Actions

BVT.13 is a potent and selective PPAR-γ activator.

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) and Nuclear Receptors (PPARs) pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Peroxisome proliferator activated receptors (PPARs) are ligand-activated transcription factors related to hormone receptors, influencing gene expression.

We offer many products related to non-steroid nuclear receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service