Skip to Content
Merck
All Photos(1)

Key Documents

B3805

Sigma-Aldrich

Anti-Bone Morphogenetic Protein 5 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-BMP-5

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 MG
CHF 437.00

CHF 437.00


Estimated to ship on03 May 2025


Request a Bulk Order

Select a Size

Change View
0.1 MG
CHF 437.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

CHF 437.00


Estimated to ship on03 May 2025


Request a Bulk Order

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

immunohistochemistry (frozen sections): 5-15 μg/mL
neutralization: suitable
western blot: 0.1 μg/mL

UniProt accession no.

application(s)

research pathology

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BMP5(653)

Immunogen

recombinant human BMP-5, expressed in NSO cells.

Biochem/physiol Actions

Bone morphogenetic proteins (BMPs) are the members of the TGF-β superfamily and are identified as the critical factors for formation of bone and cartilage. Apart from this BMPs have immune regulatory functions. The cellular effects of BMPs are mediated by a heteromeric complex composed of type I and type II receptors. BMP5 is required for the development and fine tuning of the skeletal morphology.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven Pregizer et al.
Cytokine & growth factor reviews, 20(5-6), 509-515 (2009-11-11)
Much evidence suggests that "developmental regulator" genes, like those encoding transcription factors and signaling molecules, are typically controlled by many modular, tissue-specific cis-regulatory elements that function during embryogenesis. These elements are often far from gene coding regions and promoters. Bone
Elina V García et al.
Theriogenology, 81(8), 1032-1041 (2014-03-04)
Bone morphogenetic proteins (BMPs) play a crucial role in mammalian reproduction, but little is known about their expression and function in the oviduct, where preimplantation events take place. In the present study, messenger RNA (mRNA) expression of BMPs was examined
Kohei Miyazono et al.
Cytokine & growth factor reviews, 16(3), 251-263 (2005-05-06)
Bone morphogenetic proteins (BMPs), members of the transforming growth factor-beta (TGF-beta) superfamily, bind to two different serine/threonine kinase receptors, and mediate their signals through Smad-dependent and Smad-independent pathways. Receptor regulated-Smad (R-Smad) proteins specific for the BMP pathways interact with various

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service