Skip to Content
Merck
All Photos(1)

Key Documents

B3063

Sigma-Aldrich

BI-78D3

≥98% (HPLC)

Synonym(s):

4-(2,3-Dihydrobenzol[b][1,4]dioxin-6-yl)-5-(5-nitrothiazol-2-ylthio)-4H-1,2,4-triazol-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9N5O5S2
CAS Number:
Molecular Weight:
379.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

tan

solubility

DMSO: >10 mg/mL

storage temp.

−20°C

SMILES string

Oc1nnc(Sc2ncc(s2)[N+]([O-])=O)n1-c3ccc4OCCOc4c3

InChI

1S/C13H9N5O5S2/c19-11-15-16-12(25-13-14-6-10(24-13)18(20)21)17(11)7-1-2-8-9(5-7)23-4-3-22-8/h1-2,5-6H,3-4H2,(H,15,19)

InChI key

QFRLDZGQEZCCJZ-UHFFFAOYSA-N

Biochem/physiol Actions

BI-78D3 is a substrate competitive inhibitor of JNK. JNK′s binds to JNK-interacting protein-1 (JIP1) (scaffolding protein) through high affinity D-domain on JIP1. This interaction is needed to place JNK next to target protein. BI-78D3 is a mimetic of a critical peptide structure of JIP1 which binds to JNK away from ATP binding domain preventing JIP1 JNK interaction thus acting as a substrate competitive inhibitor both in vitro and in vivo. The compound represents a growing number of modern kinase inhibitors acting at protein protein interacting areas (scaffolding) rather than ATP binding pockets.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mai Badarni et al.
JCI insight, 5 (2019-03-13)
AXL overexpression is a common resistance mechanism to anti-cancer therapies, including the resistance to BYL719 (Alpelisib) - the p110α isoform specific inhibitor of phosphoinositide 3-kinase (PI3K) - in esophagus and head and neck squamous cell carcinoma (ESCC, HNSCC respectively). However
Jianxin Zhang et al.
Experimental and therapeutic medicine, 14(6), 5445-5451 (2017-12-30)
Paeoniflorin (PAE), a principal bioactive component of
Yi Li et al.
Biomolecules & therapeutics, 26(5), 458-463 (2017-11-14)
The phosphorylation of JNK is known to induce insulin resistance in insulin target tissues. The inhibition of JNK-JIP1 interaction, which interferes JNK phosphorylation, becomes a potential target for drug development of type 2 diabetes. To discover the inhibitors of JNK-JIP1
Lu Wang et al.
Molecular and cellular endocrinology, 518, 110991-110991 (2020-08-26)
The function of the gonadotropin-releasing hormone (GnRH) neuron is critical to maintain reproductive function and a significant decrease in GnRH can lead to disorders affecting fertility, including hypogonadotropic hypogonadism. Spexin (SPX) is a novel hypothalamic neuropeptide that exerts inhibitory effects
Xiong Xiao et al.
Cell discovery, 5, 2-2 (2019-01-10)
The limited number of human hematopoietic stem cells (HSCs) has restrained their widespread clinical application. Despite great efforts in recent years, the in vitro expansion of HSCs remains a challenge due to incomplete understanding of the signaling networks underlying HSC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service