Skip to Content
Merck
All Photos(1)

Key Documents

B174

Sigma-Aldrich

BIBP 3226

Synonym(s):

N2-(Diphenylacetyl)-N-[(4-hydroxyphenyl)methyl]-D-arginine amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31N5O3
CAS Number:
Molecular Weight:
473.57
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95%

solubility

H2O: 1 mg/mL

storage temp.

−20°C

SMILES string

NC(=N)NCCC[C@@H](NC(=O)C(c1ccccc1)c2ccccc2)C(=O)NCc3ccc(O)cc3

InChI

1S/C27H31N5O3/c28-27(29)30-17-7-12-23(25(34)31-18-19-13-15-22(33)16-14-19)32-26(35)24(20-8-3-1-4-9-20)21-10-5-2-6-11-21/h1-6,8-11,13-16,23-24,33H,7,12,17-18H2,(H,31,34)(H,32,35)(H4,28,29,30)/t23-/m1/s1

InChI key

KUWBXRGRMQZCSS-HSZRJFAPSA-N

Biochem/physiol Actions

Selective non-peptide neuropeptide Y1 (NPY1) receptor antagonist.

Features and Benefits

This compound is featured on the Neuropeptide Y Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I R Tough et al.
European journal of pharmacology, 310(1), 55-60 (1996-08-22)
BIBP3226 (N2-(diphenylacetyl)-N-[4-hydroxyphenyl)methyl]-D-arginine amide) has been used to examine the presence of neuropeptide Y Y1 receptors in 3 gastrointestinal epithelial preparations, namely the rat jejunum and descending colon mucosae and a human colonic adenocarcinoma cell line. The selective Y1 receptor antagonist
Tiara Lacey et al.
Neuroscience letters, 690, 214-218 (2018-10-13)
Recent evidence indicates that Neuropeptide Y (NPY) may function as a potent anxiolytic as well as a resilience factor that can insulate the brain from the effects of stress. However, most of these studies have utilized physical stressors such as
P M Vanderheyden et al.
European journal of pharmacology, 331(2-3), 275-284 (1997-07-23)
[3H]Neuropeptide Y labelled neuropeptide Y receptors in rat forebrain membranes as a homogenous class of high-affinity sites. Between 80 and 85% of these receptors showed high affinity for Y1-selective antagonists such as (R)-N2-(diphenylacetyl)-N-[(4-hydroxyphenyl)methyl]-D-arginine amide (BIBP3226). While competitive in functional studies
Benjamin Mirman et al.
Surgery, 168(1), 155-159 (2020-06-05)
Neuropeptide Y acts directly on the vasculature as a cotransmitter with norepinephrine for an augmented contraction. Little, however, is known about the effects of neuropeptide Y on the microvasculature of human skeletal muscle. Neuropeptide Y signaling has not been studied
Olga Zagorácz et al.
Brain research, 1727, 146563-146563 (2019-11-26)
Even though several of RFamide peptides have been shown to modify memory and learning processes in different species, almost nothing is known regarding cognitive effects of recently discovered neuropeptide QRFP. Considering multiple physiological functions of QRFP, localization of QRFP-synthesizing neurons

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service