Skip to Content
Merck
All Photos(1)

Key Documents

AV48498

Sigma-Aldrich

Anti-TTC6 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC119358, Anti-MGC119360, Anti-MGC119361, Anti-Tetratricopeptide repeat domain 6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 403.00

CHF 403.00


Check Cart for Availability


Select a Size

Change View
100 μL
CHF 403.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 403.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

59 kDa

species reactivity

human, rabbit, mouse, horse, dog, bovine, pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TTC6(115669)

General description

TTC6 codes for tetratricopeptide repeat domain 6 protein. It has been identified as a susceptibility locus for allergic diseases[1].
Rabbit Anti-TTC6 antibody recognizes human, canine, and mouse TTC6.

Immunogen

Synthetic peptide directed towards the C terminal region of human TTC6

Application

Rabbit Anti-TTC6 antibody is suitable for western blot applications at a concentration of 0.5μg/ml.

Biochem/physiol Actions

The functions of TTC6 remain unknown.

Sequence

Synthetic peptide located within the following region: MKDYQDAITLNPKYSLAYFNAGNIYFHHRQFSQASDYFSKALKFDPENEY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David A Hinds et al.
Nature genetics, 45(8), 907-911 (2013-07-03)
Allergic disease is very common and carries substantial public-health burdens. We conducted a meta-analysis of genome-wide associations with self-reported cat, dust-mite and pollen allergies in 53,862 individuals. We used generalized estimating equations to model shared and allergy-specific genetic effects. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service