Skip to Content
Merck
All Photos(1)

Documents

AV47974

Sigma-Aldrich

Anti-NDRG1 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-CAP43, Anti-CMT4D, Anti-DRG1, Anti-GC4, Anti-HMSNL, Anti-N-Myc downstream Regulated gene 1, Anti-NDR1, Anti-NMSL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

43 kDa

species reactivity

human, dog, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NDRG1(10397)

General description

N-myc downstream regulated 1 (NDRG1) is a cytoplasmic hydrolase that modulates stress, hormonal responses, cell growth, differentiation and LDL receptor trafficking. It mediates antimetastatic effects in cancer cells and is downregulated in prostate cancer cells.
Rabbit Anti-NDRG1 antibody recognizes canine, chicken, mouse, zebrafish, bovine, human, and rat NDRG1.

Immunogen

Synthetic peptide directed towards the C terminal region of human NDRG1

Application

Rabbit Anti-NDRG1 antibody is suitable for western blot applications at a concentration of western blot applications at a concentration of 2.5 μg/ml.

Biochem/physiol Actions

NDRG1 is a member of the N-myc downregulated protein family which belongs to the alpha/beta hydrolase superfamily. NDRG1 is a cytoplasmic protein involved in stress responses, hormone responses, cell growth, and differentiation. Mutation in its gene has been reported to be causative for hereditary motor and sensory neuropathy-Lom.This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein involved in stress responses, hormone responses, cell growth, and differentiation. Mutation in this gene has been reported to be causative for hereditary motor and sensory neuropathy-Lom.

Sequence

Synthetic peptide located within the following region: GSSVTSLDGTRSRSHTSEGTRSRSHTSEGTRSRSHTSEGAHLDITPNSGA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Sun et al.
Carcinogenesis, 34(9), 1943-1954 (2013-05-15)
The metastasis suppressor, N-myc downstream regulated gene 1 (NDRG1), is negatively correlated with tumor progression in multiple neoplasms, being a promising new target for cancer treatment. However, the precise molecular effects of NDRG1 remain unclear. Herein, we summarize recent advances
Ranlu Liu et al.
PloS one, 8(7), e68982-e68982 (2013-07-23)
MicroRNAs, non-coding 20-22 nucleotide single-stranded RNAs, result in translational repression or degradation and gene silencing of their target genes, and significantly contribute to the regulation of gene expression. In the current study, we report that miR-182 expression was significantly upregulated
Vilja Pietiäinen et al.
Journal of cell science, 126(Pt 17), 3961-3971 (2013-07-03)
N-myc downstream-regulated gene 1 (NDRG1) mutations cause Charcot-Marie-Tooth disease type 4D (CMT4D). However, the cellular function of NDRG1 and how it causes CMT4D are poorly understood. We report that NDRG1 silencing in epithelial cells results in decreased uptake of low-density

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service