Skip to Content
Merck
All Photos(3)

Key Documents

AV46788

Sigma-Aldrich

Anti-LMAN2 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-C5orf8, Anti-GP36B, Anti-Lectin, mannose-binding 2, Anti-VIP36

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 403.00

CHF 403.00


Check Cart for Availability


Select a Size

Change View
100 μL
CHF 403.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 403.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36 kDa

species reactivity

human, bovine, rabbit, rat, mouse, dog, horse, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LMAN2(10960)

General description

Lectin, mannose-binding 2 (LMAN2, VIP36) is an intracellular, integral membrane protein that functions as a lectin of the post-Golgi secretory pathway. LMAN2 facilitates the secretion of high mannose glycoproteins predominantly via the apical surface of cells.

Specificity

Anti-LMAN2 (AB1) polyclonal antibody reacts with zebrafish, bovine, human, rat, canine, and mouse lectin, mannose-binding 2 proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human LMAN2

Application

Anti-LMAN2 (AB1) polyclonal antibody is used to tag lectin, mannose-binding 2 for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of lectin, mannose-binding 2 in the secretion of high mannose glycoproteins.

Biochem/physiol Actions

LMAN2 plays a role as an intracellular lectin in the early secretory pathway. It interacts with N-acetyl-D-galactosamine and high-mannose type glycans and may also bind to O-linked glycans. It is involved in the transport and sorting of glycoproteins carrying high mannose-type glycans.

Sequence

Synthetic peptide located within the following region: SLIKPYQGVGSSSMPLWDFQGSTMLTSQYVRLTPDERSKEGSIWNHQPCF

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arivusudar Marimuthu et al.
Proteomics. Clinical applications, 7(5-6), 355-366 (2012-11-20)
Gastric cancer is a commonly occurring cancer in Asia and one of the leading causes of cancer deaths. However, there is no reliable blood-based screening test for this cancer. Identifying proteins secreted from tumor cells could lead to the discovery

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service