Skip to Content
Merck
All Photos(1)

Key Documents

AV38677

Sigma-Aldrich

Anti-AKAP9 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-A kinase (PRKA) anchor protein (yotiao) 9

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 313.00

CHF 313.00


Check Cart for Availability


Select a Size

Change View
100 μL
CHF 313.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 313.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36 kDa

species reactivity

rat, human, guinea pig, rabbit, dog, horse, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AKAP9(10142)

Immunogen

Synthetic peptide directed towards the N terminal region of human AKAP9

Biochem/physiol Actions

A kinase (PRKA) anchor protein 9 (AKAP9) is a scaffolding protein that directs enzyme activity of protein kinases towards specific substrates. It also retains PKA and PKC in distinct subcellular compartments thus restricts the mobility of these ubiquitous enzymes with the cells.

Sequence

Synthetic peptide located within the following region: MEDEERQKKLEAGKAKLAQFRQRKAQSDGQSPSKKQKKKRKTSSSKHDVS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Terrin et al.
The Journal of cell biology, 198(4), 607-621 (2012-08-22)
Previous work has shown that the protein kinase A (PKA)-regulated phosphodiesterase (PDE) 4D3 binds to A kinase-anchoring proteins (AKAPs). One such protein, AKAP9, localizes to the centrosome. In this paper, we investigate whether a PKA-PDE4D3-AKAP9 complex can generate spatial compartmentalization
Matthew G Gold et al.
The Journal of biological chemistry, 288(24), 17111-17121 (2013-04-30)
PKA is retained within distinct subcellular environments by the association of its regulatory type II (RII) subunits with A-kinase anchoring proteins (AKAPs). Conventional reagents that universally disrupt PKA anchoring are patterned after a conserved AKAP motif. We introduce a phage

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service