Skip to Content
Merck
All Photos(1)

Key Documents

AV36000

Sigma-Aldrich

Anti-HES4 (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Hairy and enhancer of split 4 (Drosophila)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 417.00

CHF 417.00


Estimated to ship on24 May 2025



Select a Size

Change View
100 μL
CHF 417.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 417.00


Estimated to ship on24 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

24 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HES4(57801)

Immunogen

Synthetic peptide directed towards the middle region of human HES4

Biochem/physiol Actions

HES4 has been identified as transcriptional repressor that is positively regulated by Wnt signaling pathway. In conjunction with Notch signaling targets, Hes proteins regulate cell differentiation, embryonic patterning and cell fate decisions.[1][2]

Sequence

Synthetic peptide located within the following region: GHLAACLRQLGPSRRPASLSPAAPAEAPAPEVYAGRPLLPSLGGPFPLLA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kan-Ichiro Nagatomo et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 236(6), 1475-1483 (2007-04-17)
The neural crest is a population of mitotically active, multipotent progenitor cells that arise at the neural plate border. Neural crest progenitors must be maintained in a multipotent state until after neural tube closure. However, the molecular underpinnings of this
Warif El Yakoubi et al.
Stem cells (Dayton, Ohio), 30(12), 2784-2795 (2012-09-13)
The retina of fish and amphibian contains genuine neural stem cells located at the most peripheral edge of the ciliary marginal zone (CMZ). However, their cell-of-origin as well as the mechanisms that sustain their maintenance during development are presently unknown.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service