Skip to Content
Merck
All Photos(3)

Documents

AV34727

Sigma-Aldrich

Anti-KEAP1 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Kelch-like ECH-associated protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

70 kDa

species reactivity

human, rabbit, dog, rat, bovine, horse, pig, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KEAP1(9817)

General description

KEAP1 codes for a KELCH-1 like domain-containing protein that interacts with NF-E2-related factor 2. KEPAP1 has been implicated in cell signaling pathways that regulate responses to environmental and oxidative stresses.
Rabbit Anti-KEAP1 antibody recognizes pig, zebrafish, canine, bovine, human, mouse, and rat KEAP1.

Immunogen

Synthetic peptide directed towards the N terminal region of human KEAP1

Application

Rabbit Anti-KEAP1 antibody is suitable for western blot (0.5 μg/ml) applications.

Biochem/physiol Actions

KEAP1 contains KELCH-1 like domains, as well as a BTB/POZ domain. Kelch-like ECH-associated protein 1 interacts with NF-E2-related factor 2 in a redox-sensitive manner and the dissociation of the proteins in the cytoplasm is followed by transportation of NF-E2-related factor 2 to the nucleus. This interaction results in the expression of the catalytic subunit of gamma-glutamylcysteine synthetase.Western blots using two different antibodies against two unique regions of this protein target confirm the same apparent molecular weight in our tests.This gene encodes a protein containing KELCH-1 like domains, as well as a BTB/POZ domain. Kelch-like ECH-associated protein 1 interacts with NF-E2-related factor 2 in a redox-sensitive manner and the dissociation of the proteins in the cytoplasm is followed by transportation of NF-E2-related factor 2 to the nucleus. This interaction results in the expression of the catalytic subunit of gamma-glutamylcysteine synthetase. Two alternatively spliced transcript variants encoding the same isoform have been found for this gene.

Sequence

Synthetic peptide located within the following region: SQCPEGAGDAVMYASTECKAEVTPSQHGNRTFSYTLEDHTKQAFGIMNEL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas W Kensler et al.
Annual review of pharmacology and toxicology, 47, 89-116 (2006-09-14)
Keap1-Nrf2-ARE signaling plays a significant role in protecting cells from endogenous and exogenous stresses. The development of Nrf2 knockout mice has provided key insights into the toxicological importance of this pathway. These mice are more sensitive to the hepatic, pulmonary
Akira Kobayashi et al.
Molecular and cellular biology, 24(16), 7130-7139 (2004-07-30)
Transcription factor Nrf2 is a major regulator of genes encoding phase 2 detoxifying enzymes and antioxidant stress proteins in response to electrophilic agents and oxidative stress. In the absence of such stimuli, Nrf2 is inactive owing to its cytoplasmic retention
Xuedan Nie et al.
Molecular medicine reports, 19(2), 1139-1149 (2018-12-12)
The neurological disorders and neural pathology brought about by chronic alcoholism are difficult to be reversed. Increasing evidence highlights the protective roles of erythropoietin (EPO) in neurodegenerative diseases and injuries of the central nervous system. In the present study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service