AV32027
Anti-MTF1 antibody produced in rabbit
IgG fraction of antiserum
Synonym(s):
Anti-Metal-regulatory transcription factor 1
Select a Size
CHF 354.00
Select a Size
About This Item
CHF 354.00
Recommended Products
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
IgG fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
81 kDa
species reactivity
rat, human, dog, mouse, bovine, horse
concentration
0.5 mg - 1 mg/mL
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... MTF1(4520)
1 of 4
This Item | HPA028689 | HPA028604 | HPA006714 |
---|---|---|---|
antibody form IgG fraction of antiserum | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
clone polyclonal | clone polyclonal | clone polyclonal | clone polyclonal |
General description
Rabbit Anti-MTF1 antibody recognizes human, mouse, rat, bovine, and canine MTF1.
Immunogen
Application
Biochem/physiol Actions
Sequence
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service