Skip to Content
Merck
All Photos(1)

Documents

AV31648

Sigma-Aldrich

Anti-LASS3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-LAG1 homolog, ceramide synthase 3 (S. cerevisiae)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

46 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LASS3(204219)

General description

Longevity assurance homologue 3 (LASS3) is a testis-specific (dihyrdo)ceramide synthase that acts on a broad range of substrates. LASS3 is known to have two transcriptional variants, comprising of a 384-amino acid protein and a 419-amino acid protein.
Rabbit Anti-LASS3 antibody recognizes zebrafish, chicken, human, mouse, rat, bovine, and canine LASS3.

Immunogen

Synthetic peptide directed towards the N terminal region of human LASS3

Application

Rabbit Anti-LASS3 antibody can be used for western blot applications at a concentration of 0.5μg/ml.

Biochem/physiol Actions

The gene encoding the hypothetical protein LASS3 is located on chromosome 15.

Sequence

Synthetic peptide located within the following region: RVFEKFVASPLAKSFGIKETVRKVTPNTVLENFFKHSTRQPLQTDIYGLA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yukiko Mizutani et al.
The Biochemical journal, 398(3), 531-538 (2006-06-07)
The LASS (longevity assurance homologue) family members are highly conserved from yeasts to mammals. Five mouse and human LASS family members, namely LASS1, LASS2, LASS4, LASS5 and LASS6, have been identified and characterized. In the present study we cloned two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service