Skip to Content
Merck
All Photos(3)

Key Documents

AV13038

Sigma-Aldrich

Anti-GRIA2 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Glutamate receptor, ionotropic, AMPA 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 417.00

CHF 417.00


Check Cart for Availability


Select a Size

Change View
100 μL
CHF 417.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 417.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

99 kDa

species reactivity

bovine, rat, guinea pig, horse, rabbit, mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GRIA2(2891)

Immunogen

Synthetic peptide directed towards the N terminal region of human GRIA2

Application

Anti-GRIA2 (AB1) antibody produced in rabbit is suitable for western blotting at a concentration of 2.5 μg/ml.

Biochem/physiol Actions

GRIA2 or glutamate receptor 2 (GluR2) inhibits the influx of calcium through AMPA-receptor complexes. Mutations in GRIA2 gene have been associated with major psychiatric disorders such as schizophrenia and bipolar disorder.[1][2]

Sequence

Synthetic peptide located within the following region: STSEFRLTPHIDNLEVANSFAVTNAFCSQFSRGVYAIFGFYDKKSVNTIT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andy N Mead et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 23(29), 9500-9507 (2003-10-24)
Presence of the glutamate receptor 2 (GluR2) subunit prevents calcium influx through AMPA-receptor complexes; deletion of this subunit results in enhanced hippocampal long-term potentiation. We investigated whether mice lacking the GluR2 subunit [gria2 knock-out (KO) mice] displayed impairments in learning
Junyu Xu et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(46), 15415-15424 (2014-11-14)
In the CNS, synapse formation and maturation play crucial roles in the construction and consolidation of neuronal circuits. Neurexin and neuroligin localize on the opposite sides of synaptic membrane and interact with each other to promote the assembly and specialization
Alberto Chiesa et al.
European archives of psychiatry and clinical neuroscience, 262(4), 305-311 (2011-11-08)
The present study is aimed to exploring whether some single nucleotide polymorphisms (SNPs) within GRIA1, GRIA2 and GRIA4 could be associated with major depressive disorder (MDD) and whether they could predict clinical outcomes in Korean in-patients, respectively, treated with antidepressants.
Sarah L Ferri et al.
Hormones and behavior, 66(2), 409-420 (2014-07-06)
Ovarian hormones act in multiple brain regions to modulate specific behaviors and emotional states. For example, ovarian hormones promote female sexual receptivity in the hypothalamic ventromedial nucleus (VMH) and modulate anxiety in the amygdala. Hormone-induced changes within the VMH include

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service