Skip to Content
Merck
All Photos(1)

Key Documents

AV02008

Sigma-Aldrich

Anti-BCL2L1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-BCL2-like 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 324.00

CHF 324.00


Estimated to ship on30 April 2025



Select a Size

Change View
100 μL
CHF 324.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 324.00


Estimated to ship on30 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

26 kDa

species reactivity

canine, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BCL2L1(598)

General description

BCL2L1 is a signaling molecule that is known to regulate neuronal apoptosis. Furthermore, BCL2L1 can facilitate the survival of retinal ganglion cells.
Rabbit Anti-BCL2L1 associates with bovine, human, rabbit, canine, and pig BCL2L1.

Immunogen

Synthetic peptide directed towards the N terminal region of human BCL2L1

Application

Rabbit Anti-BCL2L1 can be used for western blot applications at a concentration of 1μg/ml.

Sequence

Synthetic peptide located within the following region: LVVDFLSYKLSQKGYSWSQFSDVEENRTEAPEGTESEMETPSAINGNPSW

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey M Harder et al.
Molecular and cellular neurosciences, 51(1-2), 53-59 (2012-07-28)
The Bcl-2 family is responsible for regulating cell death pathways in neurons during development, after injury and in disease. The activation of the pro-death family member BAX is often the final step before cell death in neurons. Pro-survival family members

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service