Skip to Content
Merck
All Photos(1)

Key Documents

A8168

Sigma-Aldrich

Tris(2-aminoethyl)amine

~95% (titration)

Synonym(s):

2,2′,2′′-Nitrilotriethylamine, 2,2′,2′′-Triaminotriethylamine, TAEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH2CH2CH2)3N
CAS Number:
Molecular Weight:
146.23
Beilstein:
1739626
EC Number:
MDL number:
UNSPSC Code:
12162002

vapor density

5 (vs air)

vapor pressure

0.02 mmHg ( 20 °C)

Assay

~95% (titration)

refractive index

n20/D 1.497 (lit.)

bp

114 °C/15 mmHg (lit.)

density

0.976 g/mL at 20 °C (lit.)

SMILES string

NCCN(CCN)CCN

InChI

1S/C6H18N4/c7-1-4-10(5-2-8)6-3-9/h1-9H2

InChI key

MBYLVOKEDDQJDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service