Skip to Content
Merck
All Photos(1)

Key Documents

A7009

Sigma-Aldrich

Aminoguanidine hemisulfate salt

≥98%

Synonym(s):

Guanylhydrazine hemisulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH6N4 · 1/2H2SO4
CAS Number:
Molecular Weight:
123.12
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥98%

form

powder

solubility

water: 50 mg/mL, clear to very slightly hazy, colorless

storage temp.

room temp

SMILES string

NNC(N)=N.NNC(N)=N.OS(O)(=O)=O

InChI

1S/2CH6N4.H2O4S/c2*2-1(3)5-4;1-5(2,3)4/h2*4H2,(H4,2,3,5);(H2,1,2,3,4)

InChI key

VKGQPUZNCZPZKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Aminoguanidine inhibits both constitutive and inducible nitric oxide synthetase. Aminoguanidine has also been used to study the effects of nitric oxide on ovulation and ovarian steroidogenesis and prostaglandin production.

Biochem/physiol Actions

Inhibits both constitutive and inducible nitric oxide synthetase

Features and Benefits

This compound is featured on the Nitric Oxide Synthases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Chen et al.
Oxidative medicine and cellular longevity, 2022, 8854457-8854457 (2022-01-18)
Cerebral endothelial cells play an essential role in brain angiogenesis, and their function has been found to be impaired in diabetes. Methylglyoxal (MG) is a highly reactive dicarbonyl metabolite of glucose formed mainly during glycolysis, and its levels can be
Caroline Zanotto et al.
Molecular neurobiology, 56(5), 3538-3551 (2018-08-27)
Diabetes mellitus is a metabolic disorder that results in glucotoxicity and the formation of advanced glycated end products (AGEs), which mediate several systemic adverse effects, particularly in the brain tissue. Alterations in glutamatergic neurotransmission and cognitive impairment have been reported
Fernanda Hansen et al.
Amino acids, 48(2), 375-385 (2015-09-09)
Diabetes is a metabolic disease characterized by high fasting-glucose levels. Diabetic complications have been associated with hyperglycemia and high levels of reactive compounds, such as methylglyoxal (MG) and advanced glycation endproducts (AGEs) formation derived from glucose. Diabetic patients have a
Jong Jin Park et al.
Food chemistry, 338, 128150-128150 (2020-10-24)
This study aimed to develop efficient adsorption and desorption processes to purify phenolic compounds from Ecklonia cava. We compared the adsorption and desorption properties of five resins. HP2MG showed the highest adsorption and desorption capacities and adsorption rate; hence, it
Chao Chen et al.
Nitric oxide : biology and chemistry, 70, 31-41 (2017-08-02)
Inducible nitric oxide synthase (iNOS) aggravates and endothelial nitric oxide synthase (eNOS) ameliorates fibrosis in the lung. Our previous study demonstrated that aminoguanidine (AG), a preferred iNOS inhibitor, prevents bleomycin-induced injury and fibrosis in the lung. The diethylenetriamine nitric oxide

Articles

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service