Skip to Content
Merck
All Photos(1)

Key Documents

A5229

Sigma-Aldrich

Agitoxin-1

recombinant, expressed in E. coli, ≥98% (HPLC), lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

lyophilized powder

mol wt

4021 Da

solubility

H2O: soluble
aqueous buffer: soluble

storage temp.

−20°C

Amino Acid Sequence

Gly-Val-Pro-Ile-Asn-Val-Lys-Cys-Thr-Gly-Ser-Pro-Gln-Cys-Leu-Lys-Pro-Cys-Lys-Asp-Ala-Gly-Met-Arg-Phe-Gly-Lys-Cys-Ile-Asn-Gly-Lys-Cys-His-Cys-Thr-Pro-Lys

Application

Agitoxin-1 is a potent blocker of the Shakervoltage-gated K+ channels. Agitoxin has also been used to improve multi-dimensional NMR of proteins.

Biochem/physiol Actions

Toxin found in the venom of the scorpion Leiurus quinquestriatus hebraeus. Potent blocker of the Shaker voltage-gated K+ channels as well as the mammalian homologues of Shaker

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eriks Kupce et al.
Journal of biomolecular NMR, 25(4), 349-354 (2003-05-27)
Three-dimensional HNCO and HNCA subspectra from a small protein (agitoxin, 4 kDa, enriched in carbon-13 and nitrogen-15), have been obtained by direct frequency-domain excitation of selected carbon and nitrogen sites. This new technique applies an array of several simultaneous soft
G Suarez-Kurtz et al.
The Journal of pharmacology and experimental therapeutics, 289(3), 1517-1522 (1999-05-21)
Potent and selective peptidyl blockers of the Shaker-type (Kv1) voltage-gated potassium channels were used to determine the role of these channels in regulating the spontaneous motility of smooth muscle preparations. Margatoxin (MgTX), kaliotoxin, and agitoxin-2 at 1 to 10 nM
M L Garcia et al.
Biochemistry, 33(22), 6834-6839 (1994-06-07)
Three new toxins from the venom of the scorpion Leiurus quinquestriatus var. hebraeus have been identified on the basis of their ability to block the Shaker K+ channel. These toxins have been purified using HPLC techniques and characterized as 38

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service