Skip to Content
Merck
All Photos(1)

Key Documents

A2099

Sigma-Aldrich

N-Acetyl-Leu-Glu-Val-Asp-7-amido-4-trifluoromethylcoumarin

≥95% (HPLC), solid

Synonym(s):

Ac-LEVD-AFC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H40N5O11F3
Molecular Weight:
727.68
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(C)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(O)=O)C(=O)Nc1ccc2c(OC(=O)C=C2C(F)(F)F)c1

InChI

1S/C32H40F3N5O11/c1-14(2)10-21(36-16(5)41)30(49)38-20(8-9-24(42)43)28(47)40-27(15(3)4)31(50)39-22(13-25(44)45)29(48)37-17-6-7-18-19(32(33,34)35)12-26(46)51-23(18)11-17/h6-7,11-12,14-15,20-22,27H,8-10,13H2,1-5H3,(H,36,41)(H,37,48)(H,38,49)(H,39,50)(H,40,47)(H,42,43)(H,44,45)/t20-,21-,22-,27-/m0/s1

InChI key

FETZKXHXYOHAAN-LFYAFONDSA-N

Gene Information

Amino Acid Sequence

NAc-Leu-Glu-Val-Asp-AFC

Biochem/physiol Actions

Fluorogenic substrate for caspase 4.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilona Domracheva et al.
Life sciences, 186, 92-101 (2017-08-16)
This study was designed to investigate the mechanism underlying cancer cell apoptosis caused by selenophenoquinolinones and coumarins. Twelve derivatives were studied according to their ability to suppress the proliferation of cancer cells in vitro (i.e., HepG2, MH-22A, MCF-7), induce cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service